Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnma1 Mouse anti-Mouse, HRP, Clone: S22, Abnova™

Catalog No. 89339039 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-039 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-039 Supplier Abnova Corporation Supplier No. MAB19754

Mouse Monoclonal Antibody.

Kcnma1 Monoclonal antibody specifically detects Kcnma1 in Mouse samples. It is validated for WB, IHC-P.

Specifications

Antigen Kcnma1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S22
Conjugate HRP
Description potassium large conductance calcium-activated channel, subfamily M, alpha member 1
Gene Kcnma1
Gene Alias MaxiK, Slo, Slo1, mSlo, mSlo1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 690-1196 at C-terminus of mouse Kcnma1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 16531
Target Species Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.