Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNMA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1923 Supplier Invitrogen™ Supplier No. PA1923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-923 detects large conductance, calcium-activated potassium channel alpha (Maxi K+ alpha) from human tissues as well as recombinant human protein and rat samples. PA1-923 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ~110 kDa protein representing Maxi K+ alpha from human myometrial smooth muscle cell extracts. Immunocytochemical staining of Maxi K+ alpha in human coronary muscle cells with PA1-923 results in predominant staining of the plasma membrane. PA1-923 immunizing peptide corresponds to amino acid residues 945-961 from human Maxi K+ alpha. This sequence is completely conserved across many species. PA1-923 immunizing peptide (Cat. # PEP-113) is available for use in neutralization and control experiments.

Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNMA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Kcnma1
Gene Accession No. Q12791, Q62976
Gene Alias 5730414M22Rik; bA205K10.1; big potassium channel alpha subunit; BK channel; BK channel alpha subunit; BKCa; BKCA alpha; BKCA alpha subunit; BKTM; calcium-activated potassium channel alpha subunit; calcium-activated potassium channel subunit alpha-1; calcium-activated potassium channel subunit alpha-1; uncharacterized protein; calcium-activated potassium channel, subfamily M subunit alpha-1; hSlo; K(VCA)alpha; KCa1.1; Kcnma; Kcnma1; KCNMA1b; KCNMA1c; Maxi K channel; maxiK; maxi-K channel HSLO; mSlo; mSlo1; potassium calcium-activated channel subfamily M alpha 1; potassium channel, calcium activated large conductance subfamily M alpha, member 1; potassium large conductance calcium-activated channel, subfamily M, alpha member 1; SAKCA; SLO; slo homolog; slo1; Slo-alpha; Slowpoke homolog; stretch-activated Kca channel
Gene Symbols Kcnma1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C T(945) E L V N D T N V Q F L D Q D D D(961).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3778, 83731
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.