Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNMB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106621
Description
Antibody detects endogenous levels of total KCNMB2.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.
Specifications
KCNMB2 | |
Polyclonal | |
Unconjugated | |
Kcnmb2 | |
2700049B16Rik; 3110031N04Rik; big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BKbeta2; calcium-activated potassium channel beta 2; calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; k(VCA)beta-2; Kcmb2; Kcnmb2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; potassium calcium-activated channel subfamily M regulatory beta subunit 2; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; rbeta3; Slo-beta2; Slo-beta-2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10242, 294961, 72413 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q811Q0, Q9CZM9, Q9Y691 | |
Kcnmb2 | |
A synthesized peptide derived from human KCNMB2(Accession Q9Y691), corresponding to amino acid residues S155-C205. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction