Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNMB3 Antibody (S40B-18), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212916H
Description
KCNMB3 Monoclonal antibody specifically detects KCNMB3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, MicroarraySpecifications
KCNMB3 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Microarray | |
S40B-18 | |
Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray | |
BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit | |
Fusion protein amino acids 1-49 (unique N-terminus) of mouse BKBeta3a | |
0.1 mL | |
Neuroscience | |
27094 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction