Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNMB3 Monoclonal Antibody (3E5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521690
Description
Peptide Sequence: FMLSIQREES TCTAIHTDIM DDWLDCAFTC GVHCHGQGKY PCLQVFVNLS HPGQKALLHY NEEAVQINPK CFYTPKCHQD RNDLLNSALD IKEFFDHKNG.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which may partially inactivate or slightly decrease the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 22.
Specifications
KCNMB3 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9NPA1 | |
KCNMB3 | |
KCNMB3 (NP_741979, 82 a.a. ~ 181 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
3E5 | |
Unconjugated | |
KCNMB3 | |
big potassium channel beta subunit 3; BK beta 3; BK channel beta subunit 3; BK channel subunit beta-3; BKbeta3; calcium-activated potassium channel regulatory subunit; calcium-activated potassium channel subfamily M subunit beta-3; Calcium-activated potassium channel subunit beta-3; calcium-activated potassium channel, subfamily M subunit beta-3; Charybdotoxin receptor subunit beta-3; EG435726; EMBL:BAF79924.1}; Gm5707; HBETA3; K(VCA)BETA-3; KCNMB2; KCNMB3; kcnmb3 {ECO:0000312; KCNMBL; large conductance, voltage and Ca2+ activated potassium channel Maxi K beta 3 subunit; maxi K channel subunit beta-3; MaxiK channel beta-subunit 3; potassium calcium-activated channel subfamily M regulatory beta subunit 3; potassium channel subfamily M regulatory beta subunit 3; potassium large conductance calcium-activated channel, subfamily M beta member 3; potassium large conductance calcium-activated channel, subfamily M, beta member 3; potassium large conductance calcium-activated channel, subfamily M, beta member 4; SLOBETA3; SLO-BETA-3; slobeta3 (KCNMB3) | |
Mouse | |
Affinity chromatography | |
RUO | |
27094 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction