Learn More
Invitrogen™ KCNMB4 Monoclonal Antibody (1G7)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA521735
Catalog No. PIMA521735
Only null left
Add to Cart
Description
Description
Peptide Sequence: MAKLRVAYEY TEAEDKSIRL GLFLIISGVV SLFIFGFCWL SPALQDLQAT EANCTVLSVQ QIGEVFECTF TCGADCRGTS QYPCVQVYVN NSESNSRALL HSDEHQLLTN PKCSYIPPCK RENQKNLESV MNWQQYWKDE IGSQPFTCYF NQHQRPDDVL LHRTHDEIVL LHCFLWPLVT FVVGVLIVVL TICAKSLAVK AEAMKKRKFS.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KCNMB4
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q86W47
KCNMB4
KCNMB4 (AAH50621, 1 a.a. ~ 210 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA
1G7
Unconjugated
KCNMB4
1700058G18Rik; 2900045G12Rik; big potassium channel beta subunit 4; BK channel beta subunit 4; BK channel subunit beta-4; BKbeta4; calcium activated potassium channel beta 4 subunit; calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; charybdotoxin receptor subunit beta-4; hbeta4; k(VCA)beta-4; KCMB4; KCNMB4; large conductance calcium-dependent potassium ion channel beta 4 subunit; maxi K channel subunit beta-4; MaxiK channel beta-subunit 4; potassium calcium-activated channel subfamily M regulatory beta subunit 4; potassium channel subfamily M regulatory beta subunit 4; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slo-beta-4; Slowpoke beta 4
Mouse
Affinity chromatography
RUO
27345
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.