Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNMB4 Polyclonal Antibody
KCNMB4 Polyclonal Antibody
Supplier: Thermo Scientific PA550692
Description
KCNMB4 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
KCNMB4 | |
Polyclonal | |
Unconjugated | |
KCNMB4 | |
1700058G18Rik; 2900045G12Rik; big potassium channel beta subunit 4; BK channel beta subunit 4; BK channel subunit beta-4; BKbeta4; calcium activated potassium channel beta 4 subunit; calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; charybdotoxin receptor subunit beta-4; hbeta4; k(VCA)beta-4; KCMB4; KCNMB4; large conductance calcium-dependent potassium ion channel beta 4 subunit; maxi K channel subunit beta-4; MaxiK channel beta-subunit 4; potassium calcium-activated channel subfamily M regulatory beta subunit 4; potassium channel subfamily M regulatory beta subunit 4; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slo-beta-4; Slowpoke beta 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27345, 58802 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q86W47, Q9JIN6 | |
KCNMB4 | |
Synthetic peptide corresponding to residues near the N terminal of human potassium large conductance calcium-activated channel, subfamily M, beta member 4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction