Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566810 Supplier Invitrogen™ Supplier No. PA566810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDDEEDEAGRQ RASGKPSNVG HR Highest antigen sequence identity to the following orthologs - mouse 48%, rat 48%.

Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. KCNN1 is a member of the KCNN family of potassium channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnn1
Gene Accession No. Q92952
Gene Alias AI854248; hSK1; KCa2.1; Kcnn1; potassium calcium-activated channel subfamily N member 1; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1-like; SK; Sk1; SKCa 1; SKCa1; small conductance Ca2+-activated K+ channel type1; small conductance calcium-activated potassium channel 1; small conductance calcium-activated potassium channel protein 1
Gene Symbols Kcnn1
Host Species Rabbit
Immunogen Recombinant Human KCNN1. Recombinant protein control fragment (Product #RP-107468).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.