Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114756 Supplier Thermo Scientific Supplier No. PA5114756
Add to Cart
Add to Cart

KCNN1 Polyclonal Antibody

KCNN1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Kcnn1
Gene Accession No. P70606, Q92952, Q9EQR3
Gene Alias AI854248; hSK1; KCa2.1; Kcnn1; potassium calcium-activated channel subfamily N member 1; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1-like; SK; Sk1; SKCa 1; SKCa1; small conductance Ca2+-activated K+ channel type1; small conductance calcium-activated potassium channel 1; small conductance calcium-activated potassium channel protein 1
Gene Symbols Kcnn1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNN1(Accession Q92952), corresponding to amino acid residues R390-V440.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3780, 54261, 84036
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.