Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNN4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KCNN4 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KCNN4 Polyclonal specifically detects KCNN4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KCNN4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3783 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DLVLGLGALRRRKRLLEQEKSLAG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
hIKCa1, hKCa4, hSK4, IK1, IKCa1, intermediate conductance calcium-activated potassium channel protein 4, KCa3.1IKCA1, KCa4, KCA4SKCa4, potassium intermediate/small conductance calcium-activated channel, subfamilyN, member 4, putative erythrocyte intermediate conductance calcium-activated potassiumGardos channel, Putative Gardos channel, SK4SKCa 4 | |
KCNN4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title