Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNN4 (extracellular) Monoclonal Antibody (6C1), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA527547

Catalog No. PIMA527547

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 100 uL deionized water.

Forms a voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization which promotes calcium influx. Required for maximal calcium influx and proliferation during the reactivation of naive T-cells. The channel is blocked by clotrimazole and charybdotoxin but is insensitive to apamin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCNN4 (extracellular)
Monoclonal
1 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.4
O15554, O89109, Q9QYW1
Kcnn4
Synthetic peptide mapping to the 3rd extracellular loop of human KCa 3.1
100 μg
Primary
Human, Mouse, Rat
Antibody
IgM
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
6C1
Unconjugated
KCNN4
IK1, IKCa, KCNN4
Mouse
Antigen affinity chromatography
RUO
16534, 3783, 65206
-20°C
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.