Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNN4 Mouse anti-Human, Mouse, Rat, Clone: 2A6G5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | KCNN4 |
|---|---|
| Clone | 2A6G5 |
| Concentration | 1.05 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6372
![]()
|
Proteintech Group Inc
602761IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predomit calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.Specifications
| KCNN4 | |
| 1.05 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hIKCa1, hKCa4, hSK4, IK1, IKCA1, KCa3.1, KCA4, KCNN4, Putative Gardos channel, SK4, SKCa 4, SKCa4 | |
| KCNN4 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2A6G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| O15554, O89109, Q9QYW1 | |
| 16534, 3783, 65206 | |
| KCNN4 Fusion Protein Ag19774 | |
| Primary | |
| -20°C | |
| KCNN4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title