Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144709
Description
Immunogen sequence: LQEAWMFYKH TRRKESHAAR RHQRKLLAAI NAFRQVRLKH RKLREQVNSM VDISKMHMIL YDLQQNLSSS HRALEKQIDT LAGKLDALTE LLSTALGPRQ LPEPSQQ Antigen sequence identity: Mouse 85%; Rat 82%.
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predominant calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.
Specifications
KCNN4 | |
Polyclonal | |
Unconjugated | |
Kcnn4 | |
DHS2; HIK1; hIKCa1; hKCa4; hSK4; IK; IK1; IKCa; IKCA1; intermediate conductance calcium-activated potassium channel protein 1; intermediate conductance calcium-activated potassium channel protein 4; intermediate conductance K channel; intermediate-conductance Ca-activated K channel; intermediate-conductance calcium-activated potassium channel; KCa3.1; KCA4; KCNN 4; KCNN4; mIKCa1; potassium calcium-activated channel subfamily N member 4; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 4; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4; putative erythrocyte intermediate conductance calcium-activated potassium Gardos channel; putative Gardos channel; rKCNN4c; rSK4; SK4; SKCa 4; SKCa4; SKCas; small conductance calcium-activated potassium channel 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15554 | |
Kcnn4 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 319-425. Recombinant protein control fragment (Product #RP-110219). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction