Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541015 Supplier Invitrogen™ Supplier No. PA541015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DLQQNLSSSH RALEKQIDTL AGKLDALTEL LSTALGPRQL PEPSQQSK Sequence homology: Cow: 92%; Horse: 92%; Human: 100%; Pig: 83%; Rat: 92%.

The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predominant calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNN4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kcnn4
Gene Accession No. O15554
Gene Alias DHS2; HIK1; hIKCa1; hKCa4; hSK4; IK; IK1; IKCa; IKCA1; intermediate conductance calcium-activated potassium channel protein 1; intermediate conductance calcium-activated potassium channel protein 4; intermediate conductance K channel; intermediate-conductance Ca-activated K channel; intermediate-conductance calcium-activated potassium channel; KCa3.1; KCA4; KCNN 4; KCNN4; mIKCa1; potassium calcium-activated channel subfamily N member 4; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 4; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4; putative erythrocyte intermediate conductance calcium-activated potassium Gardos channel; putative Gardos channel; rKCNN4c; rSK4; SK4; SKCa 4; SKCa4; SKCas; small conductance calcium-activated potassium channel 4
Gene Symbols Kcnn4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KCNN4 (aa 380-427).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.