Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNQ1 K+ Channel Monoclonal Antibody (S37A-10), QED Bioscience™
SDP

Catalog No. 502277431
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7431 Supplier Leinco Technologies Inc Supplier No. 11510100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCNQ1 K+ Channel Monoclonal antibody specifically detects KCNQ1 K+ Channel in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot

Specifications

Antigen KCNQ1 K+ Channel
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone S37A-10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene KCNQ1
Gene Accession No. P51787, P97414, Q9Z0N7
Gene Alias ATFB1; ATFB3; AW559127; FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; JLNS1; KCNA8; KCNA9; Kcnq1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; mutant potassium voltage-gated channel KQT-like subfamily member 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; potassium channel, voltage-gated KQT-like subfamily Q, member 1; potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel subfamily Q member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; RWS; slow delayed rectifier channel subunit; SQT2; voltage-gated potassium channel subunit Kv7.1; WRS
Host Species Mouse
Immunogen Fusion protein aa 2-101 of human KCNQ1 (K+7.1, KvLQT1, accession number P51787).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16535, 3784, 84020
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.