Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNQ1 Monoclonal Antibody (5E12)
GREENER_CHOICE

Catalog No. PIMA517105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517105 Supplier Invitrogen™ Supplier No. MA517105
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17105 targets KCNQ1 in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-17105 immunogen is purified recombinant fragment of human KCNQ1 expressed in E. Coli. MA5-17105 detects KCNQ1 which has a predicted molecular weight of approximately 95kDa.

Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. A specific K+ channel, comprised of an alpha subunit KCNQ1 and a beta subunit KCNE1, a small protein which spans the membrane only once, is predominantly expressed in the heart and in the cochlea, and is responsible for regulating the slow, depolarization-activated potassium current. Mutations in the genes encoding for KCNQ1 and KCNE1 lead to cardiac disease because they directly impair electrical signaling, and mutations in KCNQ4 are implicated in the onset of deafness. KCNQ proteins, including KCNQ1 and KCNQ4, characteristically contain six transmembrane domains and function as tetramers. KCNQ4 forms heteromeric channels with KCNQ3 and is expressed in several tissues, including the cochlea, where it is present in outer hair cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNQ1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5E12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KCNQ1
Gene Accession No. P51787
Gene Alias ATFB1; ATFB3; AW559127; FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; JLNS1; KCNA8; KCNA9; Kcnq1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; mutant potassium voltage-gated channel KQT-like subfamily member 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; potassium channel, voltage-gated KQT-like subfamily Q, member 1; potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel subfamily Q member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; RWS; slow delayed rectifier channel subunit; SQT2; voltage-gated potassium channel subunit Kv7.1; WRS
Gene Symbols KCNQ1
Host Species Mouse
Immunogen Purified recombinant fragment of human KCNQ1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3784
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.