Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNQ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144998
Description
Immunogen sequence: YSQGHLNLMV RIKELQRRLD QSIGKPSLFI SVSEKSKDRG SNTIGARLNR VEDKVTQLDQ Antigen sequence identity: Mouse 97%; Rat 97%.
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. A specific K+ channel, comprised of an alpha subunit KCNQ1 and a beta subunit KCNE1, a small protein which spans the membrane only once, is predominantly expressed in the heart and in the cochlea, and is responsible for regulating the slow, depolarization-activated potassium current. Mutations in the genes encoding for KCNQ1 and KCNE1 lead to cardiac disease because they directly impair electrical signaling, and mutations in KCNQ4 are implicated in the onset of deafness. KCNQ proteins, including KCNQ1 and KCNQ4, characteristically contain six transmembrane domains and function as tetramers. KCNQ4 forms heteromeric channels with KCNQ3 and is expressed in several tissues, including the cochlea, where it is present in outer hair cells.
Specifications
KCNQ1 | |
Polyclonal | |
Unconjugated | |
KCNQ1 | |
ATFB1; ATFB3; AW559127; FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; JLNS1; KCNA8; KCNA9; Kcnq1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; mutant potassium voltage-gated channel KQT-like subfamily member 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; potassium channel, voltage-gated KQT-like subfamily Q, member 1; potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel subfamily Q member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; RWS; slow delayed rectifier channel subunit; SQT2; voltage-gated potassium channel subunit Kv7.1; WRS | |
Rabbit | |
Affinity chromatography | |
RUO | |
3784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51787 | |
KCNQ1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 545-604. Recombinant protein control fragment (Product #RP-110230). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction