Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNQ4 K+ Channel Monoclonal Antibody (S43-6), QED Bioscience™
SDP

Catalog No. 502277432
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7432 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7432 Supplier Leinco Technologies Inc Supplier No. 11512100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCNQ4 K+ Channel Monoclonal antibody specifically detects KCNQ4 K+ Channel in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen KCNQ4 K+ Channel
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S43-6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene KCNQ4
Gene Accession No. P56696, Q9JK96, Q9JK97
Gene Alias DFNA 2; DFNA2; DFNA2A; KCNQ 4; KCNQ4; KQT like 4; KQT-like 4; KV7.4; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily KQT member 4; potassium channel KQT-like 4; potassium channel subunit alpha KvLQT4; potassium channel, voltage gated KQT-like subfamily Q, member 4; potassium channel, voltage-gated KQT-like subfamily Q, member 4; potassium voltage-gated channel KQT-like protein 4; potassium voltage-gated channel KQT-like subfamily member 4; potassium voltage-gated channel subfamily KQT member 4; potassium voltage-gated channel subfamily Q member 4; potassium voltage-gated channel, KQT-like subfamily, member 4; potassium voltage-gated channel, subfamily Q, member 4; voltage-gated potassium channel subunit Kv7.4
Host Species Mouse
Immunogen Fusion protein aa 2-77 of human KCNQ4 (K+7.4, KvLQT4, accession number P56696).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298496, 60613, 9132
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.