Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNQ4 Monoclonal Antibody (N43/6), APC
GREENER_CHOICE

Catalog No. PIMA545411
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545411 Supplier Invitrogen™ Supplier No. MA545411
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45411 was sufficient for detection of KCNQ4 in 10 μg of COS-1 cell lysate transiently expressing KCNQ4 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 77kDa.

The protein encoded by this gene forms a potassium channel that is thought to play a critical role in the regulation of neuronal excitability, particularly in sensory cells of the cochlea. The current generated by this channel is inhibited by M1 muscarinic acetylcholine receptors and activated by retigabine, a novel anti-convulsant drug. The encoded protein can form a homomultimeric potassium channel or possibly a heteromultimeric channel in association with the protein encoded by the KCNQ3 gene. Defects in this gene are a cause of nonsyndromic sensorineural deafness type 2, an autosomal dominant form of progressive hearing loss. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNQ4
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N43/6
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene KCNQ4
Gene Accession No. P56696, Q9JK96, Q9JK97
Gene Alias DFNA 2; DFNA2; DFNA2A; KCNQ 4; KCNQ4; KQT like 4; KQT-like 4; KV7.4; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily KQT member 4; potassium channel KQT-like 4; potassium channel subunit alpha KvLQT4; potassium channel, voltage gated KQT-like subfamily Q, member 4; potassium channel, voltage-gated KQT-like subfamily Q, member 4; potassium voltage-gated channel KQT-like protein 4; potassium voltage-gated channel KQT-like subfamily member 4; potassium voltage-gated channel subfamily KQT member 4; potassium voltage-gated channel subfamily Q member 4; potassium voltage-gated channel, KQT-like subfamily, member 4; potassium voltage-gated channel, subfamily Q, member 4; voltage-gated potassium channel subunit Kv7.4
Gene Symbols KCNQ4
Host Species Mouse
Immunogen Fusion protein amino acids 2-77 of human KCNQ4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298496, 60613, 9132
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.