Learn More
Invitrogen™ KCNRG Polyclonal Antibody

Description
KCNRG Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | KCNRG |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | KCNRG |
| Gene Accession No. | Q2TUM3, Q8N5I3 |
| Gene Alias | Clld4; DLTET; E030012H22Rik; Gm745; Kcnrg; Potassium channel regulator; potassium channel regulatory protein; Protein CLLD4; putative potassium channel regulatory protein |
| Gene Symbols | KCNRG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.