Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNS1 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577643
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Voltage-gated potassium channels form the largest and most diversified class of ion channels and are present in both excitable and nonexcitable cells. Their main functions are associated with the regulation of the resting membrane potential and the control of the shape and frequency of action potentials. The alpha subunits are of 2 types: those that are functional by themselves and those that are electrically silent but capable of modulating the activity of specific functional alpha subunits. The KCNS1 protein is not functional by itself but can form heteromultimers with member 1 and with member 2 (and possibly other members) of the Shab-related subfamily of potassium voltage-gated channel proteins. KCNS1 belongs to the S subfamily of the potassium channel family.
Specifications
KCNS1 (extracellular) | |
Polyclonal | |
Unconjugated | |
KCNS1 | |
KCNS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117023, 16538, 3787 | |
-20°C | |
Lyophilized |
Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O35173, O88758, Q96KK3 | |
Kcns1 | |
Peptide (C)EDVRDDPVLRRLE, corresponding to amino acid residues 233-245 of rat KV 9.1 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction