Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568672 Supplier Thermo Scientific Supplier No. PA568672
Add to Cart
Add to Cart

KCNS1 Polyclonal Antibody

KCNS1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNS1
Gene Accession No. Q96KK3
Gene Alias delayed-rectifier K(+) channel alpha subunit 1; K+ voltage-gated channel, subfamily S, 1; Kcns1; Kv9.1; potassium voltage-gated channel delayed-rectifier protein S1; potassium voltage-gated channel modifier subfamily S member 1; potassium voltage-gated channel subfamily S member 1; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 1; potassium voltage-gated channel, modifier subfamily S, member 1; voltage-gated potassium channel subunit Kv9.1
Gene Symbols KCNS1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human KCNS1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3787
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.