Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568277
Description
Predicted to react with Mouse and Rat samples.
Voltage-gated potassium channels form the largest and most diversified class of ion channels and are present in both excitable and nonexcitable cells. Their main functions are associated with the regulation of the resting membrane potential and the control of the shape and frequency of action potentials. The alpha subunits are of 2 types: those that are functional by themselves and those that are electrically silent but capable of modulating the activity of specific functional alpha subunits. The KCNS1 protein is not functional by itself but can form heteromultimers with member 1 and with member 2 (and possibly other members) of the Shab-related subfamily of potassium voltage-gated channel proteins. KCNS1 belongs to the S subfamily of the potassium channel family.
Specifications
KCNS1 | |
Polyclonal | |
Unconjugated | |
KCNS1 | |
delayed-rectifier K(+) channel alpha subunit 1; K+ voltage-gated channel, subfamily S, 1; Kcns1; Kv9.1; potassium voltage-gated channel delayed-rectifier protein S1; potassium voltage-gated channel modifier subfamily S member 1; potassium voltage-gated channel subfamily S member 1; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 1; potassium voltage-gated channel, modifier subfamily S, member 1; voltage-gated potassium channel subunit Kv9.1 | |
Rabbit | |
affinity chromatography | |
RUO | |
117023, 16538, 3787 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35173, O88758, Q96KK3 | |
KCNS1 | |
Synthesized peptide derived from human KCNS1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction