Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101769
Description
Antibody detects endogenous levels of total KCNS2.
Potassium channel subunit. Modulates channel activity and reduces the ion flow. Target protein's human homolog is a alpha subunit of a delayed-rectifier potassium channel.
Specifications
KCNS2 | |
Polyclonal | |
Unconjugated | |
KCNS2 | |
Delayed-rectifier K(+) channel alpha subunit 2; E130006J24Rik; K+ voltage-gated channel, subfamily S, 2; KCNS2; KIAA1144; Kv9.2; potassium channel, alpha subunit (kv9.2 gene); potassium voltage-gated channel modifier subfamily S member 2; potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; voltage-gated potassium channel subunit Kv9.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16539, 3788, 66022 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35174, Q9ER26, Q9ULS6 | |
KCNS2 | |
A synthesized peptide derived from human KCNS2(Accession Q9ULS6), corresponding to amino acid residues D136-S186. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction