Learn More
Invitrogen™ KCNS2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total KCNS2 protein.
Specifications
Specifications
| Antigen | KCNS2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | KCNS2 |
| Gene Accession No. | O35174, Q9ER26, Q9ULS6 |
| Gene Alias | Delayed-rectifier K(+) channel alpha subunit 2; E130006J24Rik; K+ voltage-gated channel, subfamily S, 2; KCNS2; KIAA1144; Kv9.2; potassium channel, alpha subunit (kv9.2 gene); potassium voltage-gated channel modifier subfamily S member 2; potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; voltage-gated potassium channel subunit Kv9.2 |
| Gene Symbols | KCNS2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.