Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562405
Description
Immunogen sequence: MTGQSLWDVS EANVEDGEIR INVGGFKRRL RSHTLLRF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Potassium channel subunit. Modulates channel activity and reduces the ion flow. Target protein's human homolog is a alpha subunit of a delayed-rectifier potassium channel.
Specifications
KCNS2 | |
Polyclonal | |
Unconjugated | |
KCNS2 | |
Delayed-rectifier K(+) channel alpha subunit 2; E130006J24Rik; K+ voltage-gated channel, subfamily S, 2; KCNS2; KIAA1144; Kv9.2; potassium channel, alpha subunit (kv9.2 gene); potassium voltage-gated channel modifier subfamily S member 2; potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; voltage-gated potassium channel subunit Kv9.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3788 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULS6 | |
KCNS2 | |
Recombinant protein corresponding to Human KCNS2. Recombinant protein control fragment (Product #RP-101378). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction