Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590620 Supplier Invitrogen™ Supplier No. PA590620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTGQSLWDVS EANVEDGEIR INVGGFKRRL RSHTLLRFPE TRLGRLLLCH SREAILELCD DYDDVQREFY FDRNPELFPY VLHFYHTGKL HVMAELCVFS FSQEIEYWGI NEFFIDSCCS YSYHGRKVEP EQEKWDEQSD QESTTSSFDE ILAFYNDASK FDGQPLGNFR RQLWLALDNP GYSVLSR; Positive Samples: Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein.

Potassium channel subunit. Modulates channel activity and reduces the ion flow. Target protein's human homolog is a alpha subunit of a delayed-rectifier potassium channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNS2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNS2
Gene Accession No. O35174, Q9ER26, Q9ULS6
Gene Alias Delayed-rectifier K(+) channel alpha subunit 2; E130006J24Rik; K+ voltage-gated channel, subfamily S, 2; KCNS2; KIAA1144; Kv9.2; potassium channel, alpha subunit (kv9.2 gene); potassium voltage-gated channel modifier subfamily S member 2; potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; voltage-gated potassium channel subunit Kv9.2
Gene Symbols KCNS2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16539, 3788, 66022
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.