Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576493 Supplier Invitrogen™ Supplier No. PA576493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Voltage-gated potassium channels form the largest and most diversified class of ion channels and are present in both excitable and nonexcitable cells. Their main functions are associated with the regulation of the resting membrane potential and the control of the shape and frequency of action potentials. The alpha subunits are of 2 types: those that are functional by themselves and those that are electrically silent but capable of modulating the activity of specific functional alpha subunits. The protein encoded by this gene is not functional by itself but can form heteromultimers with member 1 and with member 2 (and possibly other members) of the Shab-related subfamily of potassium voltage-gated channel proteins. This gene belongs to the S subfamily of the potassium channel family. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNS3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNS3
Gene Accession No. O88759, Q8BQZ8, Q9BQ31
Gene Alias Delayed-rectifier K(+) channel alpha subunit 3; Kcns3; KV9.3; potassium voltage-gated channel delayed-rectifier protein S3; potassium voltage-gated channel modifier subfamily S member 3; potassium voltage-gated channel subfamily S member 3; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3; potassium voltage-gated channel, modifier subfamily S, member 3; Shab-related delayed-rectifier K+ channel (Kv9.3); Shab-related delayed-rectifier K+ channel alpha subunit 3; voltage-gated potassium channel protein Kv9.3; voltage-gated potassium channel subunit Kv9.3
Gene Symbols KCNS3
Host Species Rabbit
Immunogen Recombinant protein of human KCNS3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238076, 3790, 83588
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.