Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562348 Supplier Invitrogen™ Supplier No. PA562348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIYRTESQKL TTSESQISIS VEEWEDTKDS KEQGHHRSNH RNSTSSDQSD HPLLRR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Kcnt2 calcium (Ca2+) -activated potassium channels (KCa) are a group of 6/7-TM ion channels that selectively transport K+ ions across biological membranes. KCa channels are broadly classified into three subtypes: SK, IK and BK channels (small, intermediate and big conductance). Kcnt2 is an outward rectifying potassium channel, and produces rapidly activating outward rectifier K(+) currents. Kcnt2 is activated by high intracellular sodium and chloride levels. Further, Kcnt2 channel activity is inhibited by ATP and by inhalation anesthetics, such as isoflurane. Functionally, Kcnt2 is inhibited upon stimulation of G-protein coupled receptors, such as CHRM1 and GRIA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNT2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnt2
Gene Accession No. Q6UVM3
Gene Alias E330038N15Rik; KCa4.2; Kcnt2; Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, sodium-activated subfamily T, member 2; potassium channel, subfamily T, member 2; potassium sodium-activated channel subfamily T member 2; sequence like an intermediate conductance potassium channel subunit; SLICK; SLO2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Gene Symbols Kcnt2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNT2. Recombinant protein control fragment (Product #RP-101419).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343450
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.