Learn More
Invitrogen™ KCNT2 Polyclonal Antibody

Description
KCNT2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | KCNT2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Kcnt2 |
| Gene Accession No. | Q6UVM3 |
| Gene Alias | E330038N15Rik; KCa4.2; Kcnt2; Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, sodium-activated subfamily T, member 2; potassium channel, subfamily T, member 2; potassium sodium-activated channel subfamily T member 2; sequence like an intermediate conductance potassium channel subunit; SLICK; SLO2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK) |
| Gene Symbols | Kcnt2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.