Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNT2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982009
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2009 Supplier Bioss Supplier No. BS12177R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kcnt2 calcium (Ca2+) -activated potassium channels (KCa) are a group of 6/7-TM ion channels that selectively transport K+ ions across biological membranes. KCa channels are broadly classified into three subtypes: SK, IK and BK channels (small, intermediate and big conductance). Kcnt2 is an outward rectifying potassium channel, and produces rapidly activating outward rectifier K(+) currents. Kcnt2 is activated by high intracellular sodium and chloride levels. Further, Kcnt2 channel activity is inhibited by ATP and by inhalation anesthetics, such as isoflurane. Functionally, Kcnt2 is inhibited upon stimulation of G-protein coupled receptors, such as CHRM1 and GRIA1.

Specifications

Antigen KCNT2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Kcnt2
Gene Accession No. Q6UVM3
Gene Alias E330038N15Rik; KCa4.2; KCNT2; Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, sodium-activated subfamily T, member 2; potassium channel, subfamily T, member 2; potassium sodium-activated channel subfamily T member 2; Sequence like an intermediate conductance potassium channel subunit; SLICK; SLO2.1; sodium- and chloride-activated ATP-sensitive potassium channel; Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Gene Symbols Kcnt2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KCNT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343450
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.