Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNV2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101770 Supplier Invitrogen™ Supplier No. PA5101770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNV2.

KCNV2 is a potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNV2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNV2
Gene Accession No. Q8CFS6, Q8TDN2
Gene Alias Kcnv2; KV11.1; Kv8.2; potassium channel, subfamily V, member 2; potassium channel, voltage gated modifier subfamily V, member 2; potassium channel, voltage-gated modifier subfamily V, member 2; potassium voltage-gated channel modifier subfamily V member 2; potassium voltage-gated channel subfamily V member 2; RCD3B; voltage-gated potassium channel subunit Kv8.2
Gene Symbols KCNV2
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNV2(Accession Q8TDN2), corresponding to amino acid residues M1-Q40.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169522, 240595
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.