Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNV2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578643
Description
Positive Control: 293T, A431, HeLa, HepG2 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KCNV2 is a potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.
Specifications
KCNV2 | |
Polyclonal | |
Unconjugated | |
KCNV2 | |
Kcnv2; KV11.1; Kv8.2; potassium channel, subfamily V, member 2; potassium channel, voltage gated modifier subfamily V, member 2; potassium channel, voltage-gated modifier subfamily V, member 2; potassium voltage-gated channel modifier subfamily V member 2; potassium voltage-gated channel subfamily V member 2; RCD3B; voltage-gated potassium channel subunit Kv8.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
169522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.55 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q8TDN2 | |
KCNV2 | |
Recombinant protein encompassing a sequence within the N-terminus region of human KCNV2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction