Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNV2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4038 Supplier Bioss Supplier No. BS16921R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNV2 is a potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.

Specifications

Antigen KCNV2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KCNV2
Gene Accession No. Q8TDN2
Gene Alias Kcnv2; KV11.1; Kv8.2; potassium channel, subfamily V, member 2; potassium channel, voltage gated modifier subfamily V, member 2; potassium voltage-gated channel modifier subfamily V member 2; Potassium voltage-gated channel subfamily V member 2; RCD3B; Voltage-gated potassium channel subunit Kv8.2
Gene Symbols KCNV2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KCNV2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169522
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.