Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCP Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281728F
This item is not returnable.
View return policy
Description
KCP Polyclonal antibody specifically detects KCP in Human, Mouse, Rat samples. It is validated for ELISASpecifications
KCP | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
375616 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
CRIM2, KCP1, kielin/chordin-like protein, NET67 | |
KCP antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human KCP. The immunogen is located within amino acids 1320 - 1370 of KCP. Amino Acid Squence: RAAGYRARREANAR | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction