Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564941 Supplier Invitrogen™ Supplier No. PA564941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPTVLNSHIS TPNVNALTHE NQTKPSISQI STTLPPTTST KKSGGASVVP HPSPTPLSQE EADNNEDPSI EEEDLLMLNS SPSTAKDTLD NGDYGEPDYD WTTGPRDDDE SDDTLEENRG YMEIEQSVK Highest antigen sequence identity to the following orthologs - mouse 53%, rat 56%.

C5orf15 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCT2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C5orf15
Gene Accession No. Q8NC54
Gene Alias 9530068E07Rik; C5orf15; C85061; chromosome 5 open reading frame 15; HTGN29; Kct2; keratinocyte-associated transmembrane protein 2; keratinocytes associated transmembrane protein 2; RIKEN cDNA 9530068E07 gene
Gene Symbols C5orf15
Host Species Rabbit
Immunogen Recombinant Human KCT2. Recombinant protein control fragment (Product #RP-105722).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.