Learn More
Invitrogen™ KCTD1 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGNTYILPKD SQVGPDVKSE AAPKRALYES VFGSGEICGP TSPKRLCIRP SEPVDAVVVV SVKHDPLPLL PEANGHRSTN SPT Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.
Specifications
Specifications
| Antigen | KCTD1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KCTD1 |
| Gene Accession No. | Q719H9 |
| Gene Alias | BTB/POZ domain-containing protein KCTD1; C18orf5; KCTD1; potassium channel tetramerisation domain containing 1; potassium channel tetramerization domain containing 1; potassium channel tetramerization domain-containing protein 1; SENS |
| Gene Symbols | KCTD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.