Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | KCTD10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KCTD10 Polyclonal specifically detects KCTD10 in Human samples. It is validated for Western Blot.Specifications
KCTD10 | |
Polyclonal | |
Rabbit | |
NP_114160 | |
83892 | |
Synthetic peptide directed towards the middle region of human KCTD10. Peptide sequence EETLNILLYEAQDGRGPDNALLEATGGAAGRSHHLDEDEERERIERVRRI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3, BTB/POZ domain-containing protein KCTD10, FLJ41739, MSTP028, potassium channel tetramerisation domain containing 10, Potassium channel tetramerization domain-containing protein 10, ULR061, ULRO61 | |
KCTD10 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title