Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541064
Description
Peptide sequence: MEEMSGESVV SSAVPAAATR TTSFKGTSPS SKYVKLNVGG ALYYTTMQTL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 93%.
KCTD10, a rat potassium channel tetramerisation domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.
Specifications
| KCTD10 | |
| Polyclonal | |
| Unconjugated | |
| KCTD10 | |
| AW536343; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing protein KCTD10; BTBD28; C87062; hBACURD3; KCTD10; mBACURD3; MSTP028; potassium channel tetramerisation domain containing 10; potassium channel tetramerization domain containing 10; potassium channel tetramerization domain-containing 10; potassium channel tetramerization domain-containing protein 10; ULR061; ULRO61; wu:fb30g12; zgc:63846 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 83892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H3F6 | |
| KCTD10 | |
| Synthetic peptide directed towards the N-terminal of human KCTD10 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction