Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553138 Supplier Invitrogen™ Supplier No. PA553138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEFPEARIYE ETLNILLYEA QDGRGPDNAL LEATGGAAGR SHHLDEDEER ERIERVRRIH IKRPDDRAHL HQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

KCTD10, a rat potassium channel tetramerisation domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCTD10
Gene Accession No. Q9H3F6
Gene Alias AW536343; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing protein KCTD10; BTBD28; C87062; hBACURD3; KCTD10; mBACURD3; MSTP028; potassium channel tetramerisation domain containing 10; potassium channel tetramerization domain containing 10; potassium channel tetramerization domain-containing 10; potassium channel tetramerization domain-containing protein 10; ULR061; ULRO61; wu:fb30g12; zgc:63846
Gene Symbols KCTD10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCTD10. Recombinant protein control fragment (Product #RP-90727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.