Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD11 Polyclonal Antibody

KCTD11 Polyclonal Antibody
Supplier: Thermo Scientific PA541333
Description
KCTD11 Polyclonal Antibody for Western Blot
Specifications
| KCTD11 | |
| Polyclonal | |
| Unconjugated | |
| KCTD11 | |
| AF465352; BTB/POZ domain-containing protein KCTD11; C17orf36; KCASH1; KCASH1 protein; KCTD11; potassium channel tetramerisation domain containing 11; potassium channel tetramerization domain containing 11; potassium channel tetramerization domain-containing protein 11; REN; REN/KCTD11; retinoic acid, EGF, and NGF upregulated; retinoic acid, EGF, NGF induced gene protein; retinoic acid, EGF, NGF induced gene/potassium channel tetramerization domain containing 11; RING-type E3 ubiquitin transferase KCTD11; RING-type E3 ubiquitin transferase subunit KCTD11 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 147040 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q693B1 | |
| KCTD11 | |
| Synthetic peptide directed towards the N-terminal of human KCTD11 (aa 61-110). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction