Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541088 Supplier Invitrogen™ Supplier No. PA541088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLGRLDLPRG YGETALLRAE ADFYQIRPLL DALRELEASQ GTPAPTAALL Sequence homology: Cow: 100%; Goat: 90%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCTD11
Gene Accession No. Q693B1
Gene Alias AF465352; BTB/POZ domain-containing protein KCTD11; C17orf36; KCASH1; KCASH1 protein; KCTD11; potassium channel tetramerisation domain containing 11; potassium channel tetramerization domain containing 11; potassium channel tetramerization domain-containing protein 11; REN; REN/KCTD11; retinoic acid, EGF, and NGF upregulated; retinoic acid, EGF, NGF induced gene protein; retinoic acid, EGF, NGF induced gene/potassium channel tetramerization domain containing 11; RING-type E3 ubiquitin transferase KCTD11; RING-type E3 ubiquitin transferase subunit KCTD11
Gene Symbols KCTD11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCTD11 (aa 41-90).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147040
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.