Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106417 Supplier Invitrogen™ Supplier No. PA5106417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCTD12.

KCTD12 gene ontology annotations related to this gene include protein homooligomerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kctd12
Gene Accession No. Q6WVG3, Q96CX2
Gene Alias AU046135; AW538430; BTB/POZ domain-containing protein KCTD12; C13orf2; Kctd12; KIAA1778; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein KCTD12; PFET1; Pfetin; potassium channel tetramerisation domain containing 12; potassium channel tetramerization domain containing 12; predominantly fetal expressed T1 domain; testicular tissue protein Li 100
Gene Symbols Kctd12
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCTD12(Accession Q96CX2), corresponding to amino acid residues P177-D227.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115207, 239217
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.