Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568689
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 91%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 93%.
KCTD12 gene ontology annotations related to this gene include protein homooligomerization.
Specifications
KCTD12 | |
Polyclonal | |
Unconjugated | |
Kctd12 | |
AU046135; AW538430; BTB/POZ domain-containing protein KCTD12; C13orf2; Kctd12; KIAA1778; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein KCTD12; PFET1; Pfetin; potassium channel tetramerisation domain containing 12; potassium channel tetramerization domain containing 12; predominantly fetal expressed T1 domain; testicular tissue protein Li 100 | |
Rabbit | |
Affinity chromatography | |
RUO | |
239217 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6WVG3 | |
Kctd12 | |
synthetic peptide directed towards the C-terminal region of mouse KCTD12. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction