Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541082 Supplier Invitrogen™ Supplier No. PA541082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGPAAYGLKP LTPNSKYVKL NVGGSLHYTT LRTLTGQDTM LKAMFSGRVE Sequence homology: Cow: 93%; Dog: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.

Cytochrome P450 super family proteins are monooxygenases which catalyze multiple biochemical reactions involved in drug metabolism and synthesis of cholesterol, steroids and lipids. CYP4F3, a 60KDa protein, is a novel member of this family. It consists of a conserved heme-binding domain near the C-terminus and catalyzes the u-hydroxylation of LTB4. CYP4F3 is localized in the ER and plays a vital role in the inactivation of LTB4 and controlling inflammation. CYP4F3 exists in two functionally distinct isoforms: CYP4F3A, a neutrophil isoform, is particularly expressed in neutrophils and myeloid cells and catalyzes the microsomal u-hydroxylation of LTB4. It interacts with CD11b and myeloperoxidase on myeloid cells and regulates cellular development in bone marrow. CYP4F3B, a liver isoform, catalyzes the conversion of arachidonic acid to 20-HETE, a potent activator of PKC. Itrole hasbeen implicatedin cell proliferation, vascular tone and natriuresis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD13
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCTD13
Gene Accession No. Q8WZ19
Gene Alias 1500003N18Rik; AV259508; BACURD1; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1; BTB/POZ domain-containing protein KCTD13; CTD-2574D22.4; FKSG86; hBACURD1; Kctd13; PDIP1; PDIP1alpha; Poldip1; Polymerase delta-interacting protein 1; potassium channel tetramerisation domain containing 13; potassium channel tetramerization domain containing 13; PP6832; TNFAIP1-like protein; tumor necrosis factor alpha-inducible protein PDIP1; Unknown (protein for MGC:134443)
Gene Symbols KCTD13
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCTD13 (aa 26-75).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253980
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.