Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD14 Antibody (OTI1D11), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272173MFV500
Description
KCTD14 Monoclonal antibody specifically detects KCTD14 in Human samples. It is validated for Flow CytometrySpecifications
KCTD14 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
65987 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1D11 | |
Flow Cytometry | |
BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14 | |
Full length human recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction