Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD14 Antibody (OTI1D11), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272173PECY55
Description
KCTD14 Monoclonal specifically detects KCTD14 in Human samples. It is validated for Flow Cytometry.Specifications
KCTD14 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
KCTD14 | |
Full length recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cells. | |
0.1 mL | |
65987.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1D11 | |
Flow Cytometry | |
BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction