Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD14 Antibody (OTI2E9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272171AF700
Description
KCTD14 Monoclonal specifically detects KCTD14 in Human samples. It is validated for Western Blot.Specifications
KCTD14 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KCTD14 | |
Full length recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cells. | |
0.1 mL | |
65987.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2E9 | |
Western Blot | |
BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction