Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD14 Monoclonal Antibody (OTI2B9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525488
Description
KCTD14 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
KCTD14 is a protein coding gene.
Specifications
KCTD14 | |
Monoclonal | |
0.77 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BQ13 | |
KCTD14 | |
Full length human recombinant protein of KCTD14 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2B9 | |
Unconjugated | |
KCTD14 | |
AI449310; BTB/POZ domain-containing protein KCTD14; D7Ertd760e; KCTD14; potassium channel tetramerisation domain containing 14; potassium channel tetramerization domain containing 14 | |
Mouse | |
Affinity chromatography | |
RUO | |
65987 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction