Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD14 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502662S


Description
KCTD14 Monoclonal antibody specifically detects KCTD14 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
KCTD14 | |
Monoclonal | |
0.77 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P60981, Q7M0E3 | |
Mouse | |
Affinity chromatography | |
RUO | |
11034, 502674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2B9 | |
Unconjugated | |
DSTN | |
AI449310; BTB/POZ domain-containing protein KCTD14; D7Ertd760e; KCTD14; potassium channel tetramerisation domain containing 14; potassium channel tetramerization domain containing 14 | |
Full length human recombinant protein of human DSTN produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction